Product Details

SNP ID
rs74065430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9971332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAATATATAGAAATTGAGAAAA[A/C]GGAAGAAATATGAGGAAGAAAGATG
Phenotype
MIM: 612088
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLEC12A PubMed Links

Gene Details

Gene
CLEC12A
Gene Name
C-type lectin domain family 12 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207010.1 Intron NP_001193939.1
NM_001300730.1 Intron NP_001287659.1
NM_138337.5 Intron NP_612210.4
NM_201623.3 Intron NP_963917.2
XM_005253324.2 Intron XP_005253381.1
XM_006719033.3 Intron XP_006719096.1
XM_006719035.3 Intron XP_006719098.1
XM_006719036.3 Intron XP_006719099.1
XM_011520570.1 Intron XP_011518872.1
XM_011520571.2 Intron XP_011518873.2
XM_011520573.2 Intron XP_011518875.1

View Full Product Details