Product Details

SNP ID
rs142427839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:71455798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTGTGGGGAGGGCGTAGTTCCT[A/T]ATCCCCTTTCCGGGCAGCCGCCGGG
Phenotype
MIM: 607012
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BDP1 PubMed Links

Gene Details

Gene
BDP1
Gene Name
B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018429.2 163 UTR 5 NP_060899.2
XM_011543511.2 163 UTR 5 XP_011541813.1
XM_011543512.2 163 UTR 5 XP_011541814.1
XM_017009630.1 163 UTR 5 XP_016865119.1
XM_017009631.1 163 UTR 5 XP_016865120.1
XM_017009632.1 163 UTR 5 XP_016865121.1
XM_017009633.1 163 UTR 5 XP_016865122.1
XM_017009634.1 163 UTR 5 XP_016865123.1
Gene
LOC102724392
Gene Name
uncharacterized LOC102724392
There are no transcripts associated with this gene.

View Full Product Details