Product Details

SNP ID
rs59975102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109477688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCGGCTGCTGCCCCGGGTCTGGC[A/G]GAACTCGGGTGTTTTGGGCTGAGAC
Phenotype
MIM: 613421 MIM: 608047
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD10 PubMed Links

Gene Details

Gene
KCTD10
Gene Name
potassium channel tetramerization domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317395.1 66 Intron NP_001304324.1
NM_001317399.1 66 Intron NP_001304328.1
NM_031954.4 66 Intron NP_114160.1
Gene
UBE3B
Gene Name
ubiquitin protein ligase E3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270449.1 66 UTR 5 NP_001257378.1
NM_001270450.1 66 UTR 5 NP_001257379.1
NM_001270451.1 66 UTR 5 NP_001257380.1
NM_130466.3 66 UTR 5 NP_569733.2
NM_183415.2 66 UTR 5 NP_904324.1
XM_005253987.2 66 UTR 5 XP_005254044.1
XM_006719681.3 66 UTR 5 XP_006719744.1
XM_006719682.2 66 UTR 5 XP_006719745.1
XM_011538959.2 66 UTR 5 XP_011537261.1
XM_011538961.1 66 UTR 5 XP_011537263.1
XM_017020195.1 66 UTR 5 XP_016875684.1
XM_017020196.1 66 UTR 5 XP_016875685.1

View Full Product Details