Product Details

SNP ID
rs9267810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32153817 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTTGGGGAATGCAAACTTCGTGT[C/T]CTGCTGCGGAGCCAGACGCCTGTAT
Phenotype
MIM: 603298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100507547 PubMed Links

Gene Details

Gene
LOC100507547
Gene Name
uncharacterized LOC100507547
There are no transcripts associated with this gene.

Gene
PPT2
Gene Name
palmitoyl-protein thioesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204103.1 Intron NP_001191032.1
NM_005155.6 Intron NP_005146.4
NM_138717.2 Intron NP_619731.2
Gene
PPT2-EGFL8
Gene Name
PPT2-EGFL8 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
PRRT1
Gene Name
proline rich transmembrane protein 1
There are no transcripts associated with this gene.

View Full Product Details