Product Details

SNP ID
rs4292323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:20253418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAAGCGTGCGTGGGATCAGAGG[A/C]CCGCCCTCCCCACAACAACCGGCCC
Phenotype
MIM: 603746
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLIT2 PubMed Links

Gene Details

Gene
SLIT2
Gene Name
slit guidance ligand 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289135.2 1514 UTR 5 NP_001276064.1
NM_001289136.2 1514 UTR 5 NP_001276065.1
NM_004787.3 1514 UTR 5 NP_004778.1

View Full Product Details