Product Details
- SNP ID
-
hCV27898412
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:6077606 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCGCCAGAAAATACTTTGGAGGA[A/C]CTTGGGACCCTGTGTGAGATGTATT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WSCD1
PubMed Links
Gene Details
- Gene
- WSCD1
- Gene Name
- WSC domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015253.1 |
|
Intron |
|
|
NP_056068.1 |
View Full Product Details