Product Details
- SNP ID
-
hCV27915310
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:134854886 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGTATTTATATATATTTGTCTTT[A/T]ATTTTTGGTAGAAAATGATGCATCC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
C5orf24
PubMed Links
Gene Details
- Gene
- C5orf24
- Gene Name
- chromosome 5 open reading frame 24
View Full Product Details