Product Details
- SNP ID
-
rs4426333
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:101177841 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAAGATGATTGGAAAATATATTTG[G/T]TGGCCCAGAACTGTATTTGCTCGAC
- Phenotype
-
MIM: 608183
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CHSY1
PubMed Links
Gene Details
- Gene
- CHSY1
- Gene Name
- chondroitin sulfate synthase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014918.4 |
2432 |
Missense Mutation |
CAA,CAC |
Q652H |
NP_055733.2 |
View Full Product Details