Product Details

SNP ID
rs4838921
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:111722463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGACTTGGCACTTTCAACTCTTT[A/T]CCCCTTGCCAAGAATTTGAGGTCCT
Phenotype
MIM: 179520
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM212B PubMed Links

Gene Details

Gene
FAM212B
Gene Name
family with sequence similarity 212 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019099.4 5581 UTR 3 NP_061972.1
NM_198926.2 5581 UTR 3 NP_945120.1
Gene
RAP1A
Gene Name
RAP1A, member of RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details