Product Details

SNP ID
rs6786527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150973611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAAAGAGGGATGGTGGTCAGATA[A/G]GTGACTCTATGGATTCATCATAAAA
Phenotype
MIM: 606397
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLRN1 PubMed Links

Gene Details

Gene
CLRN1
Gene Name
clarin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195794.1 Intron NP_001182723.1
NM_001256819.1 Intron NP_001243748.1
NM_052995.2 Intron NP_443721.1
NM_174878.2 Intron NP_777367.1
Gene
CLRN1-AS1
Gene Name
CLRN1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details