Product Details

SNP ID
rs6968270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:152026658 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCAGACTAAGGCACAAGTGGCTA[C/T]TCCTCTGCCTGCCCCATGGCGTAAA
Phenotype
MIM: 615130 MIM: 615133
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALNT11 PubMed Links

Gene Details

Gene
GALNT11
Gene Name
polypeptide N-acetylgalactosaminyltransferase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304514.1 Intron NP_001291443.1
NM_022087.3 Intron NP_071370.2
XM_006716082.2 Intron XP_006716145.1
XM_006716083.2 Intron XP_006716146.1
XM_006716084.2 Intron XP_006716147.1
XM_017012499.1 Intron XP_016867988.1
XM_017012500.1 Intron XP_016867989.1
Gene
GALNTL5
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 5
There are no transcripts associated with this gene.

View Full Product Details