Product Details

SNP ID
rs7696006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:71199435 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGGTATAATCTCACTACTCTGAA[A/G]AAGTTTACTTTTCCAAATAAATGCA
Phenotype
MIM: 603345
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC4A4 PubMed Links

Gene Details

Gene
SLC4A4
Gene Name
solute carrier family 4 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098484.2 Intron NP_001091954.1
NM_001134742.1 Intron NP_001128214.1
NM_003759.3 Intron NP_003750.1
XM_011532390.2 Intron XP_011530692.1
XM_017008792.1 Intron XP_016864281.1
XM_017008793.1 Intron XP_016864282.1

View Full Product Details