Product Details
- SNP ID
-
rs8117724
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45722457 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTGGGTGGTGTTAATAAAATTGC[A/G]GAGAAGATATGTGGAGACCTCAAAG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPINT4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16990631] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPINT4
- Gene Name
- serine peptidase inhibitor, Kunitz type 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178455.1 |
109 |
Silent Mutation |
GCA,GCG |
A30A |
NP_848550.1 |
- Gene
- WFDC3
- Gene Name
- WAP four-disulfide core domain 3
View Full Product Details