Product Details

SNP ID
rs6833341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:156763741 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTACACTTCAAGTCCTCTATAAAC[C/T]TGATGAGATGGATTAGATGTACTTC
Phenotype
MIM: 608452
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDGFC PubMed Links

Gene Details

Gene
PDGFC
Gene Name
platelet derived growth factor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016205.2 Intron NP_057289.1
XM_011532124.2 Intron XP_011530426.1
XM_017008455.1 Intron XP_016863944.1
XM_017008456.1 Intron XP_016863945.1

View Full Product Details