Product Details

SNP ID
rs7578228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74414385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGTTGCTCAACGGGAGGAAAGG[A/T]GCTACCTTCTTTGTCCTCTTGTTCA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C2orf81 PubMed Links

Gene Details

Gene
C2orf81
Gene Name
chromosome 2 open reading frame 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145054.1 1862 Missense Mutation ACC,TCC T571S NP_001138526.1
NM_001316764.1 1862 Missense Mutation ACC,TCC T598S NP_001303693.1
NM_001316765.1 1862 Missense Mutation ACC,TCC T549S NP_001303694.1
NM_001316766.1 1862 Missense Mutation ACC,TCC T503S NP_001303695.1
Gene
WDR54
Gene Name
WD repeat domain 54
There are no transcripts associated with this gene.

View Full Product Details