Product Details
- SNP ID
-
rs8043812
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:23651475 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGTGTATAGTTTAGGCTGACAAAC[A/G]GACTTATGTGGATGTTCTTTATTTC
- Phenotype
-
MIM: 612962
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DCTN5
PubMed Links
Gene Details
- Gene
- DCTN5
- Gene Name
- dynactin subunit 5
View Full Product Details