Product Details

SNP ID
rs6602811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15095907 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTCTAATTGTGATAAAAATCATA[C/T]TGCACTTTACCCTGTGTAACCTCCA
Phenotype
MIM: 603801 MIM: 606116
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACBD7 PubMed Links

Gene Details

Gene
ACBD7
Gene Name
acyl-CoA binding domain containing 7
There are no transcripts associated with this gene.

Gene
C10orf111
Gene Name
chromosome 10 open reading frame 111
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153244.1 1192 UTR 3 NP_694976.1
Gene
NMT2
Gene Name
N-myristoyltransferase 2
There are no transcripts associated with this gene.

Gene
RPP38
Gene Name
ribonuclease P/MRP subunit p38
There are no transcripts associated with this gene.

View Full Product Details