Product Details

SNP ID
rs6463165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:43118440 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGGTTAAGGGGGGAAAAGATGGG[A/G]AGAATAAAATTAAACATATAAGACC
Phenotype
MIM: 610384
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HECW1 PubMed Links

Gene Details

Gene
HECW1
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287059.1 Intron NP_001273988.1
NM_015052.4 Intron NP_055867.3
XM_005249665.3 Intron XP_005249722.1
XM_006715670.3 Intron XP_006715733.1
XM_006715671.3 Intron XP_006715734.1
XM_006715673.3 Intron XP_006715736.1
XM_011515220.2 Intron XP_011513522.1
XM_011515222.2 Intron XP_011513524.1
XM_011515223.2 Intron XP_011513525.1
XM_011515224.2 Intron XP_011513526.1
XM_011515225.2 Intron XP_011513527.2
XM_011515226.2 Intron XP_011513528.1
XM_017011882.1 Intron XP_016867371.1
XM_017011883.1 Intron XP_016867372.1
XM_017011884.1 Intron XP_016867373.1
XM_017011885.1 Intron XP_016867374.1
XM_017011886.1 Intron XP_016867375.1
XM_017011887.1 Intron XP_016867376.1
XM_017011888.1 Intron XP_016867377.1
XM_017011889.1 Intron XP_016867378.1
XM_017011890.1 Intron XP_016867379.1
Gene
HECW1-IT1
Gene Name
HECW1 intronic transcript 1
There are no transcripts associated with this gene.

Gene
LOC105375254
Gene Name
uncharacterized LOC105375254
There are no transcripts associated with this gene.

View Full Product Details