Product Details

SNP ID
rs5748623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:16784234 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCAGTTTCAGAGATGCAATGAA[A/C]AATGCCAGAGTCACTACACGTGAGA
Phenotype
MIM: 611674
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
XKR3 PubMed Links

Gene Details

Gene
XKR3
Gene Name
XK related 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318251.1 868 Missense Mutation TTG,TTT L255F NP_001305180.1
NM_175878.4 868 Missense Mutation TTG,TTT L255F NP_787074.2
XM_017028600.1 868 Missense Mutation TTG,TTT L255F XP_016884089.1

View Full Product Details