Product Details
- SNP ID
-
rs6873198
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:150778138 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGATTACAGAATTGGCATTGTCCTG[C/T]GTGGAAGCTGGGCCACCCACAGTGG
- Phenotype
-
MIM: 608324
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SMIM3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76036587,rs77067738] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SMIM3
- Gene Name
- small integral membrane protein 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032947.4 |
193 |
UTR 5 |
|
|
NP_116565.3 |
View Full Product Details