Product Details
- SNP ID
-
rs6994872
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:22115460 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCCCCAGTGCGGGCCTGGTACCAT[G/T]AAAAGGGGCACAGGGTGGGGGAGAT
- Phenotype
-
MIM: 602302
MIM: 615791
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74869265] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HR
- Gene Name
- hair growth associated
- Gene
- NUDT18
- Gene Name
- nudix hydrolase 18
There are no transcripts associated with this gene.
View Full Product Details