Product Details

SNP ID
rs7894175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:24589905 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCCAGTTCCATAGCAGTGCAGTT[A/G]AGGGATCCTTTTGACTAGTTTGGTT
Phenotype
MIM: 609870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP21 PubMed Links

Gene Details

Gene
ARHGAP21
Gene Name
Rho GTPase activating protein 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020824.3 Intron NP_065875.3
XM_005252542.3 Intron XP_005252599.1
XM_005252544.4 Intron XP_005252601.1
XM_011519602.2 Intron XP_011517904.1
XM_011519603.2 Intron XP_011517905.1
XM_011519604.2 Intron XP_011517906.1
XM_011519605.2 Intron XP_011517907.1
XM_011519606.2 Intron XP_011517908.1
XM_011519607.2 Intron XP_011517909.1
XM_017016459.1 Intron XP_016871948.1
XM_017016460.1 Intron XP_016871949.1
XM_017016461.1 Intron XP_016871950.1

View Full Product Details