Product Details

SNP ID
rs6468860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:103147486 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACATTGCAGATTTTTTTTTCTTCG[A/G]TTAATAGGCCCTGGTGGGAAGAACC
Phenotype
MIM: 606602
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAALC PubMed Links

Gene Details

Gene
BAALC
Gene Name
brain and acute leukemia, cytoplasmic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024372.1 Intron NP_001019543.1
NM_024812.2 Intron NP_079088.1
Gene
BAALC-AS1
Gene Name
BAALC antisense RNA 1
There are no transcripts associated with this gene.

Gene
BAALC-AS2
Gene Name
BAALC antisense RNA 2
There are no transcripts associated with this gene.

Gene
MIR3151
Gene Name
microRNA 3151
There are no transcripts associated with this gene.

View Full Product Details