Product Details

SNP ID
rs9861544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:8502734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGCCCGGAGCCCAGAAGGTCA[C/T]GGGTAAACTCCTGCACAGGAGCCTG
Phenotype
MIM: 604859
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMCD1 PubMed Links

Gene Details

Gene
LMCD1
Gene Name
LIM and cysteine rich domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278233.1 Intron NP_001265162.1
NM_001278234.1 Intron NP_001265163.1
NM_001278235.1 Intron NP_001265164.1
NM_014583.3 Intron NP_055398.1
XM_011533649.1 Intron XP_011531951.1
Gene
LMCD1-AS1
Gene Name
LMCD1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details