Product Details
- SNP ID
-
hCV29509948
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:101444935 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTATGACTACCAACAGAAGGAC[A/G]TACTGCTTGCCCTGGCTGTCTCCTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARMCX4
PubMed Links
Gene Details
- Gene
- ARMCX4
- Gene Name
- armadillo repeat containing, X-linked 4
View Full Product Details