Product Details

SNP ID
rs10512827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43367964 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGAACTGCCATGAAGTTAAGCA[A/G]TACAAATAAGGTAGGCCTGCTGGAG
Phenotype
MIM: 605240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL28 PubMed Links

Gene Details

Gene
CCL28
Gene Name
C-C motif chemokine ligand 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301873.1 Intron NP_001288802.1
NM_001301874.1 Intron NP_001288803.1
NM_001301875.1 Intron NP_001288804.1
NM_148672.3 Intron NP_683513.1

View Full Product Details