Product Details

SNP ID
rs10466409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13392333 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAGGGGAGTGCAAGGTAGGCAGA[C/G]AGAGTATCATTCTAAGGAGAGAATG
Phenotype
MIM: 602550 MIM: 615933
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARNTL PubMed Links

Gene Details

Gene
ARNTL
Gene Name
aryl hydrocarbon receptor nuclear translocator like
There are no transcripts associated with this gene.

Gene
BTBD10
Gene Name
BTB domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297741.1 Intron NP_001284670.1
NM_001297742.1 Intron NP_001284671.1
NM_032320.6 Intron NP_115696.2
XM_017018405.1 Intron XP_016873894.1
XM_017018406.1 Intron XP_016873895.1
XM_017018407.1 Intron XP_016873896.1
XM_017018408.1 Intron XP_016873897.1
XM_017018409.1 Intron XP_016873898.1
XM_017018410.1 Intron XP_016873899.1
XM_017018411.1 Intron XP_016873900.1

View Full Product Details