Product Details

SNP ID
rs10413463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50196247 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGAGATGAGGGAACCCTTAACAC[A/G]CTTTGAACCTTGGGAAGTTCACCTG
Phenotype
MIM: 608568
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYH14 PubMed Links

Gene Details

Gene
MYH14
Gene Name
myosin, heavy chain 14, non-muscle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077186.1 Intron NP_001070654.1
NM_001145809.1 Intron NP_001139281.1
NM_024729.3 Intron NP_079005.3
XM_006723386.3 Intron XP_006723449.1
XM_011527320.2 Intron XP_011525622.1
XM_011527321.2 Intron XP_011525623.1
XM_011527323.2 Intron XP_011525625.1

View Full Product Details