Product Details

SNP ID
rs5991935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101418955 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCAAGGACAGACAGCTCACCAACC[A/G]TGACAGCACCATACTGGAACTTCAA
Phenotype
MIM: 300610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMCX4 PubMed Links

Gene Details

Gene
ARMCX4
Gene Name
armadillo repeat containing, X-linked 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256155.2 Intron NP_001243084.2
Gene
HNRNPH2
Gene Name
heterogeneous nuclear ribonucleoprotein H2 (H')
There are no transcripts associated with this gene.

Gene
RPL36A-HNRNPH2
Gene Name
RPL36A-HNRNPH2 readthrough
There are no transcripts associated with this gene.

View Full Product Details