Product Details
- SNP ID
-
hCV29715797
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:117088806 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGTATGAAGACAAGAACAAATGG[C/G]AGTGTCCTTTTATACCACTCACTCC
- Phenotype
-
MIM: 600833
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ST7
PubMed Links
Gene Details
- Gene
- ST7
- Gene Name
- suppression of tumorigenicity 7
There are no transcripts associated with this gene.
- Gene
- ST7-AS2
- Gene Name
- ST7 antisense RNA 2
There are no transcripts associated with this gene.
View Full Product Details