Product Details

SNP ID
rs9881425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:123491817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATACAAAAATGACTATATATA[C/T]GGTTGTACAATTTTTTACCCAAATT
Phenotype
MIM: 615939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HACD2 PubMed Links

Gene Details

Gene
HACD2
Gene Name
3-hydroxyacyl-CoA dehydratase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017005855.1 3667 UTR 3 XP_016861344.1
XM_017005856.1 3667 Intron XP_016861345.1

View Full Product Details