Product Details

SNP ID
rs9827382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:127195395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAAGGCTCCCTGCCATGCTCGC[C/T]CTGAGAGTTCTGGCAGGCACTCCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C3orf56 PubMed Links

Gene Details

Gene
C3orf56
Gene Name
chromosome 3 open reading frame 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007534.2 Intron NP_001007535.1

View Full Product Details