Product Details

SNP ID
rs10399772
Assay Type
Functionally Tested
NCBI dbSNP Submissions
12
Location
Chr.1:225488012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACGTTGTTCTTTAACCAGAAAG[A/G]TTCACTGCTGACTCCCCCACCTCTC
Phenotype
MIM: 609061
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ENAH PubMed Links

Gene Details

Gene
ENAH
Gene Name
enabled homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008493.1 11758 UTR 3 NP_001008493.1
NM_018212.4 11758 UTR 3 NP_060682.2
XM_005273187.2 11758 UTR 3 XP_005273244.1
XM_011544229.2 11758 Intron XP_011542531.2
XM_017001746.1 11758 UTR 3 XP_016857235.1
XM_017001747.1 11758 Intron XP_016857236.1
XM_017001748.1 11758 Intron XP_016857237.1
XM_017001749.1 11758 Intron XP_016857238.1
XM_017001750.1 11758 Intron XP_016857239.1
XM_017001751.1 11758 UTR 3 XP_016857240.1
XM_017001752.1 11758 Intron XP_016857241.1
XM_017001753.1 11758 UTR 3 XP_016857242.1
XM_017001754.1 11758 UTR 3 XP_016857243.1
XM_017001755.1 11758 UTR 3 XP_016857244.1
XM_017001756.1 11758 UTR 3 XP_016857245.1
XM_017001757.1 11758 UTR 3 XP_016857246.1

View Full Product Details