Product Details
- SNP ID
-
rs4772975
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:108498311 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTGAAGGCGAAGAGGTCCAGAAGA[C/G]TTAGGTAATTTTTTTTTCAAGATCA
- Phenotype
-
MIM: 615479
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MYO16
PubMed Links
Gene Details
- Gene
- MYO16
- Gene Name
- myosin XVI
View Full Product Details