Product Details

SNP ID
rs9493515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:133095017 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATACAAAATGAATAAGACAAATT[C/T]ATAGCATCTCTGGGCAGGGAAAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00326 PubMed Links

Gene Details

Gene
LINC00326
Gene Name
long intergenic non-protein coding RNA 326
There are no transcripts associated with this gene.

View Full Product Details