Product Details

SNP ID
rs9818032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:115804497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCTCACTTTAAAGTTGTCAGTT[T/G]ATCTTAGATGATGAGGAAGCAGTAC
Phenotype
MIM: 603241
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
LSAMP PubMed Links

Gene Details

Gene
LSAMP
Gene Name
limbic system-associated membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318915.1 7344 UTR 3 NP_001305844.1
NM_002338.4 7344 UTR 3 NP_002329.2
XM_011512840.2 7344 Intron XP_011511142.1
XM_017006383.1 7344 Intron XP_016861872.1
XM_017006384.1 7344 Intron XP_016861873.1

View Full Product Details