Product Details

SNP ID
rs9304616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42874755 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATACGCCTGTCAGGAAGCTAGAA[C/G]TCTCTAGAAAGTGACCGGAGAATGT
Phenotype
MIM: 176390
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PSG1 PubMed Links

Gene Details

Gene
PSG1
Gene Name
pregnancy specific beta-1-glycoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184825.1 Intron NP_001171754.1
NM_001184826.1 Intron NP_001171755.1
NM_001297773.1 Intron NP_001284702.1
NM_006905.2 Intron NP_008836.2
XM_005259065.3 Intron XP_005259122.1
XM_011527125.1 Intron XP_011525427.1
XM_017026994.1 Intron XP_016882483.1

View Full Product Details