Product Details

SNP ID
rs10164880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169597388 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTCACTGCAGCCTCAAAATCCTC[A/G]GCTGAAAGAGTCCTCCCACCTGATC
Phenotype
MIM: 606093
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPIG PubMed Links

Gene Details

Gene
PPIG
Gene Name
peptidylprolyl isomerase G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004792.2 Intron NP_004783.2
XM_005246966.1 Intron XP_005247023.1
XM_005246967.1 Intron XP_005247024.1
XM_017005302.1 Intron XP_016860791.1

View Full Product Details