Product Details

SNP ID
rs9490937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:123810481 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAACTATGAGCAGCTGGTGTATG[C/G]ACTAGAGAATAAAAGGGCAGAAGGG
Phenotype
MIM: 609758
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NKAIN2 PubMed Links

Gene Details

Gene
NKAIN2
Gene Name
Na+/K+ transporting ATPase interacting 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040214.2 Intron NP_001035304.1
NM_001300737.1 Intron NP_001287666.1
NM_001300738.1 Intron NP_001287667.1
NM_001300740.1 Intron NP_001287669.1
NM_153355.4 Intron NP_699186.2
XM_011535501.2 Intron XP_011533803.1
XM_011535502.2 Intron XP_011533804.1
XM_011535503.2 Intron XP_011533805.1
XM_017010318.1 Intron XP_016865807.1
XM_017010319.1 Intron XP_016865808.1

View Full Product Details