Product Details

SNP ID
rs6084306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3212971 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAGCAGAGACCTGACGTAGAAGA[A/G]ATAGAGAAGCAAGGAGATGGAAGGG
Phenotype
MIM: 616177 MIM: 147520
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDRGK1 PubMed Links

Gene Details

Gene
DDRGK1
Gene Name
DDRGK domain containing 1
There are no transcripts associated with this gene.

Gene
ITPA
Gene Name
inosine triphosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267623.1 Intron NP_001254552.1
NM_001324236.1 Intron NP_001311165.1
NM_001324237.1 Intron NP_001311166.1
NM_001324238.1 Intron NP_001311167.1
NM_001324240.1 Intron NP_001311169.1
NM_033453.3 Intron NP_258412.1
NM_181493.3 Intron NP_852470.1
XM_006723564.3 Intron XP_006723627.1
XM_006723565.3 Intron XP_006723628.1
XM_011529234.2 Intron XP_011527536.1

View Full Product Details