Product Details
- SNP ID
-
rs6062384
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:62956346 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGGAGCACACACCCGGGGTGCAG[C/T]CAGCTCCCCGCAGGGGCAGACAGGA
- Phenotype
-
MIM: 611625
MIM: 612107
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GID8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113393446] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GID8
- Gene Name
- GID complex subunit 8 homolog
There are no transcripts associated with this gene.
- Gene
- SLC17A9
- Gene Name
- solute carrier family 17 member 9
View Full Product Details