Product Details

SNP ID
rs7526293
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:154471733 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTGTGTGTAAGCATTCACACTA[C/T]AGGTGTATGCATTATTAAATTAAGG
Phenotype
MIM: 147880 MIM: 610482
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL6R PubMed Links

Gene Details

Gene
IL6R
Gene Name
interleukin 6 receptor
There are no transcripts associated with this gene.

Gene
SHE
Gene Name
Src homology 2 domain containing E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010846.2 Intron NP_001010846.1
XM_005244891.4 Intron XP_005244948.1
XM_011509163.2 Intron XP_011507465.1
XM_017000256.1 Intron XP_016855745.1

View Full Product Details