Product Details

SNP ID
rs5933426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:134576106 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTTGAACTTACATCACTGGACTA[A/C]AAATGTTTAGGAATACCTATCTATG
Phenotype
MIM: 300296
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLAC1 PubMed Links

Gene Details

Gene
PLAC1
Gene Name
placenta specific 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316887.1 Intron NP_001303816.1
NM_001316888.1 Intron NP_001303817.1
NM_001316889.1 Intron NP_001303818.1
NM_021796.3 Intron NP_068568.1
XM_011531257.2 Intron XP_011529559.1
XM_011531260.2 Intron XP_011529562.1
XM_017029236.1 Intron XP_016884725.1

View Full Product Details