Product Details

SNP ID
rs6096076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50743001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTTAAGAGTATTAGAGTTTATC[C/T]TAATTGATTCGGGAGATGGGGAAGA
Phenotype
MIM: 608975
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARD6B PubMed Links

Gene Details

Gene
PARD6B
Gene Name
par-6 family cell polarity regulator beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032521.2 Intron NP_115910.1

View Full Product Details