Product Details

SNP ID
rs6700036
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:100084814 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAAGGCTTAATTCATTTCAGTTA[A/T]TTAAGGTAAATTTAGGACTTTTCCC
Phenotype
MIM: 609321
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MFSD14A PubMed Links

Gene Details

Gene
MFSD14A
Gene Name
major facilitator superfamily domain containing 14A
There are no transcripts associated with this gene.

Gene
SASS6
Gene Name
SAS-6 centriolar assembly protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304829.1 2689 UTR 3 NP_001291758.1
NM_194292.2 2689 UTR 3 NP_919268.1
XM_017000486.1 2689 UTR 3 XP_016855975.1

View Full Product Details