Product Details

SNP ID
rs6551253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:28371475 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCTTACTAAAGGCCAAGGAGTTA[C/T]ACATCAAAGGAAGTTGAACAAATAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZCWPW2 PubMed Links

Gene Details

Gene
ZCWPW2
Gene Name
zinc finger CW-type and PWWP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040432.3 Intron NP_001035522.1
NM_001324169.1 Intron NP_001311098.1
NM_001324170.1 Intron NP_001311099.1
XM_017005755.1 Intron XP_016861244.1
XM_017005756.1 Intron XP_016861245.1
XM_017005757.1 Intron XP_016861246.1
XM_017005758.1 Intron XP_016861247.1
XM_017005759.1 Intron XP_016861248.1

View Full Product Details