Product Details

SNP ID
rs9982910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30829504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGAAGCTACCACCATAGCAGCC[G/T]CCCAGGTTGTTGATGTTGCTACCAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP7-1 PubMed Links

Gene Details

Gene
KRTAP7-1
Gene Name
keratin associated protein 7-1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181606.2 229 Silent Mutation NP_853637.2

View Full Product Details