Product Details

SNP ID
rs6103934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44943214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGCAAAGCTGGATTTATAGTC[C/T]TAATAGAAGAAAAGACAGGAGTGTG
Phenotype
MIM: 616049
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PABPC1L PubMed Links

Gene Details

Gene
PABPC1L
Gene Name
poly(A) binding protein cytoplasmic 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001124756.2 Intron NP_001118228.1
Gene
TOMM34
Gene Name
translocase of outer mitochondrial membrane 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006809.4 Intron NP_006800.2

View Full Product Details