Product Details

SNP ID
rs7534953
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:177176004 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTTATGCTTCAAACTGGGGACTA[C/T]GGTGAACCAATCTCGTCTGACCAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRINP2 PubMed Links

Gene Details

Gene
BRINP2
Gene Name
BMP/retinoic acid inducible neural specific 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021165.3 Intron NP_066988.1
XM_005245379.1 Intron XP_005245436.1
XM_011509826.1 Intron XP_011508128.1

View Full Product Details