Product Details
- SNP ID
-
rs9542034
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:69701204 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAATGAGGCAGAAGCAGGTTTGTTT[G/T]AATAAAGCATAATTCTGCTTCCCGT
- Phenotype
-
MIM: 605332
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs41283970] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLHL1
- Gene Name
- kelch like family member 1
View Full Product Details